Learn More
Abcam Native Human Apolipoprotein CII/ApoC-II, 50UG
Catalog No. 502709065
Encompass_Preferred
Native Human Apolipoprotein CII/ApoC-II is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: APC2, APOC2, Apolipoprotein C-II, Apo-CII, ApoC-II, Apolipoprotein C2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9065 Supplier Abcam Supplier No. AB9092550UG
Native Human Apolipoprotein CII/ApoC-II is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: APC2, APOC2, Apolipoprotein C-II, Apo-CII, ApoC-II, Apolipoprotein C2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement