Learn More
Abcam Native human Carboxypeptidase B2 protein, 50UG
Catalog No. 502709598
Encompass_Preferred
Native human Carboxypeptidase B2 protein is a Human Full Length protein, in the 23 to 423 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Carboxypeptidase B2, Carboxypeptidase U, Plasma carboxypeptidase B, Thrombin-activable fibrinolysis inhibitor, CPU, pCPB, TAFI, CPB2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9598 Supplier Abcam Supplier No. AB8174950UG
Native human Carboxypeptidase B2 protein is a Human Full Length protein, in the 23 to 423 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Carboxypeptidase B2, Carboxypeptidase U, Plasma carboxypeptidase B, Thrombin-activable fibrinolysis inhibitor, CPU, pCPB, TAFI, CPB2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement