Learn More
Abcam Native human Cathepsin G protein, 100UG
Catalog No. 502709049
Encompass_Preferred
Native human Cathepsin G protein is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Cathepsin G, CG, CTSG
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9049 Supplier Abcam Supplier No. AB91122100UG
Native human Cathepsin G protein is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Cathepsin G, CG, CTSG
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement