Learn More
Creative Biomart Native Human CEACAM5, 100ug | Purity : > 98%
Catalog No. NC3662482
Encompass
Native Human Carcinoembryonic Antigen was purified from Human Liver Metastases.Members of the CEACAM subfamily, including CEACAM5, belong to the CEA gene family. For general information on the CEA gene family, see CEACAM1.
Catalog No. NC3662482 Supplier Creative Biomart Supplier No. CEACAM527803TH
Native Human Carcinoembryonic Antigen was purified from Human Liver Metastases.Members of the CEACAM subfamily, including CEACAM5, belong to the CEA gene family. For general information on the CEA gene family, see CEACAM1.