Learn More
Abcam Native Human Collagen I protein, 500UG
Catalog No. 502709828
Encompass_Preferred
Native Human Collagen I protein is a Human Full Length protein, expressed in Native, with >95% purity and suitable for WB, SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Collagen alpha-1(I) chain, Alpha-1 type I collagen, COL1A1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9828 Supplier Abcam Supplier No. AB7533500UG
Native Human Collagen I protein is a Human Full Length protein, expressed in Native, with >95% purity and suitable for WB, SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Collagen alpha-1(I) chain, Alpha-1 type I collagen, COL1A1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement