Learn More
Abcam Native Human Factor Va protein, 50UG
Catalog No. 502710240
Encompass_Preferred
Native Human Factor Va protein is a Human Full Length protein with >95% purity and suitable for Functional studies and SDS-PAGE. The predicted molecular weight of ab62415 protein is 168 kDa (based on combined MW of subunits). - Save time and ensure accurate results- use our Factor Va protein as a control - Two subunits- Heavy chain: 94 kDa and light chain: 74 kDa - Optimal protein bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor V, Activated protein C cofactor, F5
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-0240 Supplier Abcam Supplier No. AB6241550UG
Native Human Factor Va protein is a Human Full Length protein with >95% purity and suitable for Functional studies and SDS-PAGE. The predicted molecular weight of ab62415 protein is 168 kDa (based on combined MW of subunits). - Save time and ensure accurate results- use our Factor Va protein as a control - Two subunits- Heavy chain: 94 kDa and light chain: 74 kDa - Optimal protein bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor V, Activated protein C cofactor, F5
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement