Learn More
Abcam Native human Factor Xa, 100UG
Catalog No. 502710258
Encompass_Preferred
Native human Factor Xa is a Human Full Length protein with >95% purity and suitable for SDS-PAGE and Functional studies. The predicted molecular weight of ab62229 native protein is 46 kDa. - Save time and ensure accurate results - use our Factor Xa protein as a control - Optimal protein bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor X, Stuart factor, Stuart-Prower factor, F10
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-0258 Supplier Abcam Supplier No. AB62229100UG
Native human Factor Xa is a Human Full Length protein with >95% purity and suitable for SDS-PAGE and Functional studies. The predicted molecular weight of ab62229 native protein is 46 kDa. - Save time and ensure accurate results - use our Factor Xa protein as a control - Optimal protein bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor X, Stuart factor, Stuart-Prower factor, F10
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement