Learn More
Abcam Native Human Factor XIII protein (Active), 100UG
Catalog No. 502710236
Encompass_Preferred
Native Human Factor XIII protein (Active) is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor XIII B chain, Fibrin-stabilizing factor B subunit, Protein-glutamine gamma-glutamyltransferase B chain, Transglutaminase B chain, F13B
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-0236 Supplier Abcam Supplier No. AB62427100UG
Native Human Factor XIII protein (Active) is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor XIII B chain, Fibrin-stabilizing factor B subunit, Protein-glutamine gamma-glutamyltransferase B chain, Transglutaminase B chain, F13B
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement