Learn More
Abcam Native Human IgA protein, 1MG
Catalog No. NC2190986
Encompass_Preferred
Native Human IgA protein is a Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab91025 native protein is 160 kDa. - Save time and ensure accurate results- use our native IgA protein as an isotype control - No cross-reaction from other immunoglobulin isotypes
Protein/ Peptide/ Enzyme Synonyms: IGHA2, IGHA1, IGHA, Ig alpha 1 chain C region, Ig alpha 2 chain C region
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
This item is not returnable. View return policy
Catalog No. NC2190986 Supplier Abcam Supplier No. AB910251MG
Native Human IgA protein is a Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab91025 native protein is 160 kDa. - Save time and ensure accurate results- use our native IgA protein as an isotype control - No cross-reaction from other immunoglobulin isotypes
Protein/ Peptide/ Enzyme Synonyms: IGHA2, IGHA1, IGHA, Ig alpha 1 chain C region, Ig alpha 2 chain C region
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement