Learn More
Abcam Native Human IgA1 protein, 1MG
Catalog No. NC2028155
Encompass_Preferred
Native Human IgA1 protein is a Human Full Length IgA1 protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab91020 native protein is 160 kDa. - Save time and ensure accurate results- use our native IgA1 protein as an isotype control - No cross-reaction from other immunoglobulin isotypes
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant alpha 1, Ig alpha-1 chain C region, Ig alpha-1 chain C region BUR, Ig alpha-1 chain C region TRO, IGHA1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2028155 Supplier Abcam Supplier No. AB910201MG
Native Human IgA1 protein is a Human Full Length IgA1 protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab91020 native protein is 160 kDa. - Save time and ensure accurate results- use our native IgA1 protein as an isotype control - No cross-reaction from other immunoglobulin isotypes
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant alpha 1, Ig alpha-1 chain C region, Ig alpha-1 chain C region BUR, Ig alpha-1 chain C region TRO, IGHA1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement