Learn More
Abcam Native Human IgG3 protein, 1MG
Catalog No. NC2070095
Encompass_Preferred
Native Human IgG3 protein is a Human Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab118426 native protein is 170 kDa. - Save time and ensure accurate results - use our IgG3 protein as an isotype control - No cross-reaction from other immunoglobulin isotypes
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant gamma 3, HDC, Heavy chain disease protein, Ig gamma-3 chain C region, IGHG3
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2070095 Supplier Abcam Supplier No. AB1184261MG
Native Human IgG3 protein is a Human Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab118426 native protein is 170 kDa. - Save time and ensure accurate results - use our IgG3 protein as an isotype control - No cross-reaction from other immunoglobulin isotypes
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant gamma 3, HDC, Heavy chain disease protein, Ig gamma-3 chain C region, IGHG3
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement