Learn More
Abcam Native Human IgM Fc fragment protein, 1MG
Catalog No. 502709101
Encompass_Preferred
Native Human IgM Fc fragment protein is a Human Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab90287 native protein is 342 kDa. - Save time and ensure accurate results- use our IgM Fc fragment protein as a control - Optimal protein bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant mu, Ig mu chain C region, Ig mu chain C region BOT, Ig mu chain C region GAL, Ig mu chain C region OU, IGHM
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9101 Supplier Abcam Supplier No. AB902871MG
Native Human IgM Fc fragment protein is a Human Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab90287 native protein is 342 kDa. - Save time and ensure accurate results- use our IgM Fc fragment protein as a control - Optimal protein bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant mu, Ig mu chain C region, Ig mu chain C region BOT, Ig mu chain C region GAL, Ig mu chain C region OU, IGHM
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement