Learn More
Abcam Native human LDL protein, 1MG
Catalog No. 502709050
Encompass_Preferred
Native human LDL protein is a Full Length protein with >95% purity and suitable for Functional studies and SDS-PAGE. The predicted molecular weight of ab91115 native protein is 2300 kDa. - Save time and ensure accurate results - use our Low density lipoprotein (LDL) as a control
Protein/ Peptide/ Enzyme Synonyms: Low density lipoprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9050 Supplier Abcam Supplier No. AB911151MG
Native human LDL protein is a Full Length protein with >95% purity and suitable for Functional studies and SDS-PAGE. The predicted molecular weight of ab91115 native protein is 2300 kDa. - Save time and ensure accurate results - use our Low density lipoprotein (LDL) as a control
Protein/ Peptide/ Enzyme Synonyms: Low density lipoprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement