Learn More
Abcam Native human PF4 protein, 100UG
Catalog No. 502709594
Encompass_Preferred
Native human PF4 protein is a Human Full Length Platelet Factor 4 protein, in the 32 to 101 aa range with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab81754 native protein is 7.8 kDa. - Save time and ensure accurate results - use our PF4 protein - Heparin-neutralizing activity verified by clotting assay- Marker of platelet activation- Optimal protein bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: CXCL4, SCYB4, PF4, Platelet factor 4, PF-4, C-X-C motif chemokine 4, Iroplact, Oncostatin-A
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9594 Supplier Abcam Supplier No. AB81754100UG
Native human PF4 protein is a Human Full Length Platelet Factor 4 protein, in the 32 to 101 aa range with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab81754 native protein is 7.8 kDa. - Save time and ensure accurate results - use our PF4 protein - Heparin-neutralizing activity verified by clotting assay- Marker of platelet activation- Optimal protein bioactivity and stability
Protein/ Peptide/ Enzyme Synonyms: CXCL4, SCYB4, PF4, Platelet factor 4, PF-4, C-X-C motif chemokine 4, Iroplact, Oncostatin-A
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement