Learn More
Abcam Native Human Von Willebrand Factor protein, 100UG
Catalog No. 502709192
Encompass_Preferred
Native Human Von Willebrand Factor protein is a Human Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab88533 native protein is 260 kDa. - Save time and ensure accurate results- use our Von Willebrand Factor protein as a control.
Protein/ Peptide/ Enzyme Synonyms: F8VWF, VWF, von Willebrand factor, vWF
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9192 Supplier Abcam Supplier No. AB88533100UG
Native Human Von Willebrand Factor protein is a Human Full Length protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab88533 native protein is 260 kDa. - Save time and ensure accurate results- use our Von Willebrand Factor protein as a control.
Protein/ Peptide/ Enzyme Synonyms: F8VWF, VWF, von Willebrand factor, vWF
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement