Learn More
Abcam Native mouse Factor X protein, 100UG
Catalog No. 502708391
Encompass_Preferred
Native mouse Factor X protein is a Mouse Full Length protein, in the 41 to 481 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor X, Stuart factor, F10
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-8391 Supplier Abcam Supplier No. AB96742100UG
Native mouse Factor X protein is a Mouse Full Length protein, in the 41 to 481 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor X, Stuart factor, F10
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement