Learn More
Abcam Native Mouse Fibrinogen protein (Active), 1MG
Supplier:  Abcam AB927911MG
Encompass_Preferred
Native Mouse Fibrinogen protein (Active) is a Mouse Full Length protein, in the 1 to 481 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Fibrinogen gamma chain, Fgg
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2036162