Learn More
Apexbio Technology LLC Native-PAGE Gel Preparation and Electrophoresis Kit (PI ≤ 7.0) 50tests
SDP
Catalog No. 5000097031
Encompass_Preferred
Native-PAGE Gel Preparation and Electrophoresis Kit PI le 7 0 is a reagent kit for the separation and analysis of functional proteins under non-denaturing conditions It is designed to enable separation of proteins based on intrinsic charge and molecular conformation while preserving native activity and structure Native-PAGE Gel Preparation and Electrophoresis Kit PI le 7 0 utilizes a Tris-Glycine buffer system (25 mM Tris 192 mM Glycine) for electrophoretic separation facilitating the migration of acidic proteins (pI 7 0) toward the anode under slightly basic conditions This kit does not require denaturants such as SDS or -mercaptoethanol Based on these properties Native-PAGE Gel Preparation and Electrophoresis Kit PI le 7 0 is suitable for research applications in the characterization and purification of biologically active macromolecules including isoenzyme and protein complex identification
Catalog No. 50-000-97031 Supplier Apexbio Technology LLC Supplier No. K414250
Native-PAGE Gel Preparation and Electrophoresis Kit PI le 7 0 is a reagent kit for the separation and analysis of functional proteins under non-denaturing conditions It is designed to enable separation of proteins based on intrinsic charge and molecular conformation while preserving native activity and structure Native-PAGE Gel Preparation and Electrophoresis Kit PI le 7 0 utilizes a Tris-Glycine buffer system (25 mM Tris 192 mM Glycine) for electrophoretic separation facilitating the migration of acidic proteins (pI 7 0) toward the anode under slightly basic conditions This kit does not require denaturants such as SDS or -mercaptoethanol Based on these properties Native-PAGE Gel Preparation and Electrophoresis Kit PI le 7 0 is suitable for research applications in the characterization and purification of biologically active macromolecules including isoenzyme and protein complex identification