Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp PFAS Standard Solution, Liquid, 1.2 mL, >98% Purity

Catalog No. 502392428 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This standard solution contains eleven native linear perfluoroalkylcarboxylic acids (C4-C14), seven native perfluoroalkylsulfonates, three native telomer sulfonates, two native linear and branched perfluorooctanesulfonamidoacetic acids, three native perfluoroalkylsulfonamides, sodium dodecafluoro-3H-4,8-dioxanonanoate, major/minor components of F-53B, and GenX. Designed for PFAS separation and review, it assesses isomer-specific quantitation. Individual native components have over 98% purity, commonly used in PFAS detection.

  • Liquid formulation
  • 1.2 mL volume
  • One unit per package
  • Reversed phase separation
  • LCMS detection
  • LC/MS system compatible
  • Quantitative analysis
  • Ambient shipping
  • Components with >98% purity
  • Hazardous material, special freight

Catalog No. 50-239-2428 Supplier Waters Corp Supplier No. PFAC30PAR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This standard solution contains eleven native linear perfluoroalkylcarboxylic acids (C4-C14), seven native perfluoroalkylsulfonates, three native telomer sulfonates, two native linear and branched perfluorooctanesulfonamidoacetic acids, three native perfluoroalkylsulfonamides, sodium dodecafluoro-3H-4,8-dioxanonanoate, major/minor components of F-53B, and GenX. Designed for PFAS separation and review, it assesses isomer-specific quantitation. Individual native components have over 98% purity, commonly used in PFAS detection.

  • Liquid formulation
  • 1.2 mL volume
  • One unit per package
  • Reversed phase separation
  • LCMS detection
  • LC/MS system compatible
  • Quantitative analysis
  • Ambient shipping
  • Components with >98% purity
  • Hazardous material, special freight

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.