Learn More
Creative Biomart Native Porcine Mucin protein, 100g
Supplier:  Creative Biomart MUCIN357
Encompass
Mucin played an important role in many functions.Source :Porcine Gastric MucosaSpecies :PorcineForm :White to yellow powderPurity :Partially Purified. Mucin: 60.0 - 90.0%Quality Control Test :Ash: 3.0 - 5.5%Mucin Nitrogen: 7.0 - 10.0%Reducing Substances: < 45%Salmonella: Negative;E. coli: Negative;Loss on Drying: < 6.0%;pH: 3.0 - 5.0 (2% solution);Storage :2-8 centigrade
Catalog No. NC1734991