Learn More
MOLECULAR DEPOT LLC NativeFolder Medium - 1 L
Catalog No. 5000013441
Encompass_Preferred
NativeFolder Bacterial Culture Medium (1L) is a unique bacterial culture medium formulated to promote the folding and the expression of soluble recombinant proteins in E coli NativeFolder contains proprietary compounds that prevent protein aggregation and misfolding during protein expression This product is supplied as two powdered mixes to be reconstituted with di-water and combined to produce 1L of bacterial culture medium Large bulk volumes are available at large discounts For in vitro research use only
Catalog No. 50-000-13441 Supplier MOLECULAR DEPOT LLC Supplier No. K1010004
NativeFolder Bacterial Culture Medium (1L) is a unique bacterial culture medium formulated to promote the folding and the expression of soluble recombinant proteins in E coli NativeFolder contains proprietary compounds that prevent protein aggregation and misfolding during protein expression This product is supplied as two powdered mixes to be reconstituted with di-water and combined to produce 1L of bacterial culture medium Large bulk volumes are available at large discounts For in vitro research use only