Learn More
GENOSKIN NativeSkin access 8mm - Ex vivo human skin model
Catalog No. NC1927956
Encompass
Ready-to-use human skin model with dedicated culture medium – ext. diam. 8mm. NativeSkin is a live, patented ex vivo human skin model made from round biopsies of donated surgical discards. Preserving the natural structure and functionality of real human skin, it is maintained in a living state for 7 days using Genoskin's patented technology. NativeSkin supports topical and systemic-like studies, making it ideal for researching transdermal delivery, skin metabolism, and other skin-related effects. This model provides precise, non-clinical testing with highly relevant human data, offering an ethical alternative to animal testing.
Catalog No. NC1927956 Supplier GENOSKIN Supplier No. NSA08
Ready-to-use human skin model with dedicated culture medium – ext. diam. 8mm. NativeSkin is a live, patented ex vivo human skin model made from round biopsies of donated surgical discards. Preserving the natural structure and functionality of real human skin, it is maintained in a living state for 7 days using Genoskin's patented technology. NativeSkin supports topical and systemic-like studies, making it ideal for researching transdermal delivery, skin metabolism, and other skin-related effects. This model provides precise, non-clinical testing with highly relevant human data, offering an ethical alternative to animal testing.