Learn More
MULTISORB NATRASORB Silica Gel Desiccant, Prevents Moisture Damage, Non-Dusting Solution, 1 Unit
SDP
Catalog No. NC1620046
Encompass_Preferred
This item has a minimum order quantity of 4 cases per supplier requirements.
The MULTISORB NATRASORB 1U T S 1300 is a military-grade silica gel desiccant packet designed for effective moisture control in various applications including pharmaceuticals and electronics. With a capacity of 2.85 g at 20% RH and 5.70 g at 40% RH, it prevents rust, corrosion, and inhibits mold, enhancing product shelf life. Its non-dusting and puncture-proof Tyvek material ensures reliable performance, setting it apart from conventional desiccants.
Catalog No. NC1620046 Supplier MULTISORB Supplier No. 0203127DG01
This item has a minimum order quantity of 4 cases per supplier requirements.
The MULTISORB NATRASORB 1U T S 1300 is a military-grade silica gel desiccant packet designed for effective moisture control in various applications including pharmaceuticals and electronics. With a capacity of 2.85 g at 20% RH and 5.70 g at 40% RH, it prevents rust, corrosion, and inhibits mold, enhancing product shelf life. Its non-dusting and puncture-proof Tyvek material ensures reliable performance, setting it apart from conventional desiccants.