Learn More
Hampton Research NATRIX 2 NO02 185ML
SDP
Catalog No. 501657
Encompass
HR2-917-02/Natrix 2 #02 (E02), 185 ml - 0.08 M Sodium chloride, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 5.5, 35% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.002 M Hexammine cobalt(III) chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-165-7 Supplier Hampton Research Supplier No. HR291702
HR2-917-02/Natrix 2 #02 (E02), 185 ml - 0.08 M Sodium chloride, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 5.5, 35% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.002 M Hexammine cobalt(III) chloride. Please note items from this supplier are non-returnable.