Learn More
Hampton Research NATRIX 2 NO03 185ML
SDP
Catalog No. 501658
Encompass
HR2-917-03/Natrix 2 #03 (E03), 185 ml - 0.012 M Sodium chloride, 0.08 M Potassium chloride, 0.04 M Sodium cacodylate trihydrate pH 5.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.002 M Hexammine cobalt(III) chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-165-8 Supplier Hampton Research Supplier No. HR291703
HR2-917-03/Natrix 2 #03 (E03), 185 ml - 0.012 M Sodium chloride, 0.08 M Potassium chloride, 0.04 M Sodium cacodylate trihydrate pH 5.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.002 M Hexammine cobalt(III) chloride. Please note items from this supplier are non-returnable.