Learn More
Hampton Research NATRIX 2 NO04 185ML
SDP
Catalog No. 501659
Encompass
HR2-917-04/Natrix 2 #04 (E04), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 5.5, 40% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.002 M Hexammine cobalt(III) chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-165-9 Supplier Hampton Research Supplier No. HR291704
HR2-917-04/Natrix 2 #04 (E04), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 5.5, 40% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.002 M Hexammine cobalt(III) chloride. Please note items from this supplier are non-returnable.