Learn More
Hampton Research NATRIX 2 NO05 185ML
SDP
Catalog No. 501660
Encompass
HR2-917-05/Natrix 2 #05 (E05), 185 ml - 0.002 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 1.8 M Ammonium sulfate, 0.0005 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-166-0 Supplier Hampton Research Supplier No. HR291705
HR2-917-05/Natrix 2 #05 (E05), 185 ml - 0.002 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 1.8 M Ammonium sulfate, 0.0005 M Spermine. Please note items from this supplier are non-returnable.