Learn More
Hampton Research NATRIX 2 NO06 185ML
SDP
Catalog No. 501661
Encompass
HR2-917-06/Natrix 2 #06 (E06), 185 ml - 0.05 M Sodium cacodylate trihydrate pH 6.0, 35% v/v Tacsimate pH 6.0, 0.001 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-166-1 Supplier Hampton Research Supplier No. HR291706
HR2-917-06/Natrix 2 #06 (E06), 185 ml - 0.05 M Sodium cacodylate trihydrate pH 6.0, 35% v/v Tacsimate pH 6.0, 0.001 M Spermine. Please note items from this supplier are non-returnable.