Learn More
Hampton Research NATRIX 2 NO07 185ML
SDP
Catalog No. 501662
Encompass
HR2-917-07/Natrix 2 #07 (E07), 185 ml - 0.1 M Sodium chloride, 0.05 M Sodium cacodylate trihydrate pH 6.0, 10% w/v Polyethylene glycol 4,000, 0.0005 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-166-2 Supplier Hampton Research Supplier No. HR291707
HR2-917-07/Natrix 2 #07 (E07), 185 ml - 0.1 M Sodium chloride, 0.05 M Sodium cacodylate trihydrate pH 6.0, 10% w/v Polyethylene glycol 4,000, 0.0005 M Spermine. Please note items from this supplier are non-returnable.