Learn More
Hampton Research NATRIX 2 NO08 185ML
SDP
Catalog No. 501663
Encompass
HR2-917-08/Natrix 2 #08 (E08), 185 ml - 0.05 M Potassium chloride, 0.05 M Sodium cacodylate trihydrate pH 6.0, 10% w/v Polyethylene glycol 8,000, 0.0005 M Spermine, 0.0005 M L-Argininamide dihydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-166-3 Supplier Hampton Research Supplier No. HR291708
HR2-917-08/Natrix 2 #08 (E08), 185 ml - 0.05 M Potassium chloride, 0.05 M Sodium cacodylate trihydrate pH 6.0, 10% w/v Polyethylene glycol 8,000, 0.0005 M Spermine, 0.0005 M L-Argininamide dihydrochloride. Please note items from this supplier are non-returnable.