Learn More
Hampton Research NATRIX 2 NO09 185ML
SDP
Catalog No. 501664
Encompass
HR2-917-09/Natrix 2 #09 (E09), 185 ml - 0.1 M Potassium chloride, 0.05 M Sodium cacodylate trihydrate pH 6.0, 16% w/v Polyethylene glycol 1,000, 0.0005 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-166-4 Supplier Hampton Research Supplier No. HR291709
HR2-917-09/Natrix 2 #09 (E09), 185 ml - 0.1 M Potassium chloride, 0.05 M Sodium cacodylate trihydrate pH 6.0, 16% w/v Polyethylene glycol 1,000, 0.0005 M Spermine. Please note items from this supplier are non-returnable.