Learn More
Hampton Research NATRIX 2 NO10 185ML
SDP
Catalog No. 501665
Encompass
HR2-917-10/Natrix 2 #10 (E10), 185 ml - 0.005 M Magnesium chloride hexahydrate, 0.002 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 15% v/v 2-Propanol, 0.001 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-166-5 Supplier Hampton Research Supplier No. HR291710
HR2-917-10/Natrix 2 #10 (E10), 185 ml - 0.005 M Magnesium chloride hexahydrate, 0.002 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 15% v/v 2-Propanol, 0.001 M Spermine. Please note items from this supplier are non-returnable.