Learn More
Hampton Research NATRIX 2 NO14 185ML
SDP
Catalog No. 501669
Encompass
HR2-917-14/Natrix 2 #14 (F02), 185 ml - 0.08 M Sodium chloride, 0.012 M Potassium chloride, 0.02 M Magnesium chloride, 0.04 M Sodium cacodylate pH 6.0, 30% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-166-9 Supplier Hampton Research Supplier No. HR291714
HR2-917-14/Natrix 2 #14 (F02), 185 ml - 0.08 M Sodium chloride, 0.012 M Potassium chloride, 0.02 M Magnesium chloride, 0.04 M Sodium cacodylate pH 6.0, 30% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.