Learn More
Hampton Research NATRIX 2 NO15 185ML
SDP
Catalog No. 501670
Encompass
HR2-917-15/Natrix 2 #15 (F03), 185 ml - 0.08 M Sodium chloride, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 6.0, 35% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-167-0 Supplier Hampton Research Supplier No. HR291715
HR2-917-15/Natrix 2 #15 (F03), 185 ml - 0.08 M Sodium chloride, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 6.0, 35% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.