Learn More
Hampton Research NATRIX 2 NO16 185ML
SDP
Catalog No. 501671
Encompass
HR2-917-16/Natrix 2 #16 (F04), 185 ml - 0.08 M Strontium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 6.0, 35% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-167-1 Supplier Hampton Research Supplier No. HR291716
HR2-917-16/Natrix 2 #16 (F04), 185 ml - 0.08 M Strontium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 6.0, 35% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.