Learn More
Hampton Research NATRIX 2 NO18 185ML
SDP
Catalog No. 501673
Encompass
HR2-917-18/Natrix 2 #18 (F06), 185 ml - 0.08 M Potassium chloride, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 6.0, 45% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-167-3 Supplier Hampton Research Supplier No. HR291718
HR2-917-18/Natrix 2 #18 (F06), 185 ml - 0.08 M Potassium chloride, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 6.0, 45% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.