Learn More
Hampton Research NATRIX 2 NO19 185ML
SDP
Catalog No. 501674
Encompass
HR2-917-19/Natrix 2 #19 (F07), 185 ml - 0.08 M Sodium chloride, 0.04 M Sodium cacodylate trihydrate pH 6.0, 45% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-167-4 Supplier Hampton Research Supplier No. HR291719
HR2-917-19/Natrix 2 #19 (F07), 185 ml - 0.08 M Sodium chloride, 0.04 M Sodium cacodylate trihydrate pH 6.0, 45% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.