Learn More
Hampton Research NATRIX 2 NO20 185ML
SDP
Catalog No. 501675
Encompass
HR2-917-20/Natrix 2 #20 (F08), 185 ml - 0.08 M Sodium chloride, 0.02 M Barium chloride dihydrate, 0.04 M Sodium cacodylate trihydrate pH 6.0, 45% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-167-5 Supplier Hampton Research Supplier No. HR291720
HR2-917-20/Natrix 2 #20 (F08), 185 ml - 0.08 M Sodium chloride, 0.02 M Barium chloride dihydrate, 0.04 M Sodium cacodylate trihydrate pH 6.0, 45% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.