Learn More
Hampton Research NATRIX 2 NO23 185ML
SDP
Catalog No. 501678
Encompass
HR2-917-23/Natrix 2 #23 (F11), 185 ml - 0.018 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 10% v/v 2-Propanol, 0.003 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-167-8 Supplier Hampton Research Supplier No. HR291723
HR2-917-23/Natrix 2 #23 (F11), 185 ml - 0.018 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.5, 10% v/v 2-Propanol, 0.003 M Spermine. Please note items from this supplier are non-returnable.