Learn More
Hampton Research NATRIX 2 NO24 185ML
SDP
Catalog No. 501679
Encompass
HR2-917-24/Natrix 2 #24 (F12), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M MOPS pH 7.0, 2.0 M Ammonium sulfate, 0.0005 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-167-9 Supplier Hampton Research Supplier No. HR291724
HR2-917-24/Natrix 2 #24 (F12), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M MOPS pH 7.0, 2.0 M Ammonium sulfate, 0.0005 M Spermine. Please note items from this supplier are non-returnable.