Learn More
Hampton Research NATRIX 2 NO26 185ML
SDP
Catalog No. 501681
Encompass
HR2-917-26/Natrix 2 #26 (G02), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M MOPS pH 7.0, 55% v/v Tacsimate pH 7.0, 0.002 M Hexammine cobalt(III) chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-168-1 Supplier Hampton Research Supplier No. HR291726
HR2-917-26/Natrix 2 #26 (G02), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M MOPS pH 7.0, 55% v/v Tacsimate pH 7.0, 0.002 M Hexammine cobalt(III) chloride. Please note items from this supplier are non-returnable.