Learn More
Hampton Research NATRIX 2 NO27 185ML
SDP
Catalog No. 501682
Encompass
HR2-917-27/Natrix 2 #27 (G03), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 7.0, 15% v/v 2-Propanol, 0.001 M Hexammine cobalt(III) chloride, 0.001 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-168-2 Supplier Hampton Research Supplier No. HR291727
HR2-917-27/Natrix 2 #27 (G03), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 7.0, 15% v/v 2-Propanol, 0.001 M Hexammine cobalt(III) chloride, 0.001 M Spermine. Please note items from this supplier are non-returnable.