Learn More
Hampton Research NATRIX 2 NO28 185ML
SDP
Catalog No. 501683
Encompass
HR2-917-28/Natrix 2 #28 (G04), 185 ml - 0.005 M Magnesium chloride hexahydrate, 0.05 M MOPS pH 7.0, 25% v/v 1,4-Dioxane, 0.001 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-168-3 Supplier Hampton Research Supplier No. HR291728
HR2-917-28/Natrix 2 #28 (G04), 185 ml - 0.005 M Magnesium chloride hexahydrate, 0.05 M MOPS pH 7.0, 25% v/v 1,4-Dioxane, 0.001 M Spermine. Please note items from this supplier are non-returnable.