Learn More
Hampton Research NATRIX 2 NO29 185ML
SDP
Catalog No. 501684
Encompass
HR2-917-29/Natrix 2 #29 (G05), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.002 M Barium chloride dihydrate, 0.05 M MOPS pH 7.0, 30% v/v 1,4-Dioxane. Please note items from this supplier are non-returnable.
Catalog No. 50-168-4 Supplier Hampton Research Supplier No. HR291729
HR2-917-29/Natrix 2 #29 (G05), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.002 M Barium chloride dihydrate, 0.05 M MOPS pH 7.0, 30% v/v 1,4-Dioxane. Please note items from this supplier are non-returnable.