Learn More
Hampton Research NATRIX 2 NO30 185ML
SDP
Catalog No. 501685
Encompass
HR2-917-30/Natrix 2 #30 (G06), 185 ml - 0.001 M Magnesium chloride hexahydrate, 0.002 M Calcium chloride dihydrate, 0.05 M MOPS pH 7.0, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-168-5 Supplier Hampton Research Supplier No. HR291730
HR2-917-30/Natrix 2 #30 (G06), 185 ml - 0.001 M Magnesium chloride hexahydrate, 0.002 M Calcium chloride dihydrate, 0.05 M MOPS pH 7.0, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.