Learn More
Hampton Research NATRIX 2 NO31 185ML
SDP
Catalog No. 501686
Encompass
HR2-917-31/Natrix 2 #31 (G07), 185 ml - 0.08 M Strontium chloride hexahydrate, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 7.0, 20% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-168-6 Supplier Hampton Research Supplier No. HR291731
HR2-917-31/Natrix 2 #31 (G07), 185 ml - 0.08 M Strontium chloride hexahydrate, 0.02 M Magnesium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 7.0, 20% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.