Learn More
Hampton Research NATRIX 2 NO32 185ML
SDP
Supplier:  Hampton Research HR291732
Encompass
HR2-917-32/Natrix 2 #32 (G08), 185 ml - 0.08 M Sodium chloride, 0.04 M Sodium cacodylate trihydrate pH 7.0, 30% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-168-7