Learn More
Hampton Research NATRIX 2 NO33 185ML
SDP
Catalog No. 501688
Encompass
HR2-917-33/Natrix 2 #33 (G09), 185 ml - 0.04 M Lithium chloride, 0.08 M Strontium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 7.0, 30% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-168-8 Supplier Hampton Research Supplier No. HR291733
HR2-917-33/Natrix 2 #33 (G09), 185 ml - 0.04 M Lithium chloride, 0.08 M Strontium chloride hexahydrate, 0.04 M Sodium cacodylate trihydrate pH 7.0, 30% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.