Learn More
Hampton Research NATRIX 2 NO34 185ML
SDP
Supplier:  Hampton Research HR291734
Encompass
HR2-917-34/Natrix 2 #34 (G10), 185 ml - 0.04 M Lithium chloride, 0.08 M Strontium chloride, 0.02 M Magnesium chloride, 0.04 M Sodium cacodylate pH 7.0, 30% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-168-9