Learn More
Hampton Research NATRIX 2 NO37 185ML
SDP
Catalog No. 501692
Encompass
HR2-917-37/Natrix 2 #37 (H01), 185 ml - 0.08 M Sodium chloride, 0.02 M Barium chloride dihydrate, 0.04 M Sodium cacodylate trihydrate pH 7.0, 40% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-169-2 Supplier Hampton Research Supplier No. HR291737
HR2-917-37/Natrix 2 #37 (H01), 185 ml - 0.08 M Sodium chloride, 0.02 M Barium chloride dihydrate, 0.04 M Sodium cacodylate trihydrate pH 7.0, 40% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.012 M Spermine tetrahydrochloride. Please note items from this supplier are non-returnable.